Learn More
Medchemexpress LLC 2-chloro-4-nitropyridine | 23056-36-2 | MFCD00006316 | 99.9% | 158.54 | 10 G
SDP
Catalog No. 5000453515
Encompass_Preferred
2-Chloro-4-nitropyridine is a chemical compound primarily utilized as a drug intermediate in various synthetic processes. It appears as a white to yellow solid.
  • Used in synthesizing other organic compounds.
  • Intermediate for 2-chloro-4-ethoxypyridine.
  • Intermediate for various RET kinase inhibitors.
Catalog No. 50-004-53515 Supplier Medchemexpress LLC Supplier No. HYW01017410G
Add to Cart
Add to Cart
2-Chloro-4-nitropyridine is a chemical compound primarily utilized as a drug intermediate in various synthetic processes. It appears as a white to yellow solid.
  • Used in synthesizing other organic compounds.
  • Intermediate for 2-chloro-4-ethoxypyridine.
  • Intermediate for various RET kinase inhibitors.