Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. C22831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22831G 1 g
C22835G 5 g
C228325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C22831G Supplier TCI America Supplier No. C22831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23056-36-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00661454
InChI Key LIEPVGBDUYKPLC-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-4-nitro, 2-chloro-4-nitro-pyridine, 2-chlor-4-nitropyridin, pubchem9220, 2-chloro-4nitropyridine, 2chloro-4-nitropyridine, acmc-1ccv7, 2-chloro-4-nitro pyridine, ksc201q1f, 2-chloro-4-nitropyridine
PubChem CID 735152
IUPAC Name 2-chloro-4-nitropyridine
SMILES [O-][N+](=O)C1=CC(Cl)=NC=C1

Specifications

Melting Point 54°C
Quantity 1 g
Formula Weight 158.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.