Learn More
Accela Chembio Inc 2-chloro-4-pyridylboronic Acid | 25g | 458532-96-2 | MFCD03788416 | 97+% | Shelf Life: 1440 Days | Light Sensitive/n2/+4
Supplier:  Accela Chembio Inc SY00603725G
Encompass_Preferred
2-chloro-4-pyridylboronic Acid | 25g | 458532-96-2 | MFCD03788416 | 97+% | Shelf Life: 1440 Days | Light Sensitive/n2/+4
Catalog No. 50-354-440