Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C17901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17901G 1 g
C17905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17901G Supplier TCI America Supplier No. C17901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175205-54-6
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.05
MDL Number MFCD00052912
InChI Key NJXDBSSSDPOAFI-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
PubChem CID 2736675
IUPAC Name 2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
SMILES FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 279.05
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.