Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-(trifluoromethyl)benzonitrile 95.0+%, TCI America™
SDP

Catalog No. C25995G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25995G Supplier TCI America Supplier No. C25995G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1813-33-8
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
MDL Number MFCD00084939
InChI Key GEHMLBFNZKJDQM-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzonitrile, 3-chloro-4-cyanobenzotrifluoride, 3-chloro-4-cyano-benzotrifluoride, 4-trifluoromethyl-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl benzenecarbonitrile, pubchem4745, acmc-209eho, buttpark 9757-31, ksc494o6f
PubChem CID 2736476
IUPAC Name 2-chloro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)C#N

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
UN Number 3276
Formula Weight 205.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.