Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluoro-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. C31441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31441G 1 g
C31445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31441G Supplier TCI America Supplier No. C31441G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38186-84-4
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD04972725
InChI Key FMPYGEFGXUAAKG-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-3-picoline, 2-chloro-3-methyl-5-fluoropyridine, 2-chloro-5-fluoro-3-methyl-pyridine, pyridine, 2-chloro-5-fluoro-3-methyl, pubchem6220, 5-fluoro-2-chloro-3-methylpyridine, 5-fluoro-2-chloro-3-methyl-pyridine, 2-chloro-3-methy-5-fluorolpyridine; 2-chloro-5-fluoro-3-picoline
PubChem CID 12852014
IUPAC Name 2-chloro-5-fluoro-3-methylpyridine
SMILES CC1=CC(=CN=C1Cl)F

Specifications

Melting Point 43°C
Color White
Quantity 1 g
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-5-fluoro-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.