Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorotoluene 97.0+%, TCI America™
SDP

Catalog No. C15375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15375G 5 g
C153725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15375G Supplier TCI America Supplier No. C15375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33406-96-1
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00040853
InChI Key NIOGDCDTKPQEAT-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorotoluene, 3-fluoro-6-chlorotoluene, benzene, 1-chloro-4-fluoro-2-methyl, 1-chlor-4-fluor-2-methylbenzol, pubchem1606, acmc-1ajiz, ksc493m2j, 6-chloro-3-fluorotoluene
PubChem CID 118475
IUPAC Name 1-chloro-4-fluoro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)F)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 144.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.