Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-hydroxybenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. C15161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15161G 1 g
C15165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15161G Supplier TCI America Supplier No. C15161G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6294-93-5
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.55
MDL Number MFCD00042525
InChI Key ZLFPIEUWXNRPNM-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenol, 2-chloro-5-hydroxybenzotrifluoride, phenol, 4-chloro-3-trifluoromethyl, 4-chloro-3-trifluoromethyl-phenol, phenol, p-chloro-m-trifluoromethyl, p-chloro-m-trifluoromethylphenol, 2-chloro-5-hydroxybezotrifluoride, pubchem2863
PubChem CID 80520
IUPAC Name 4-chloro-3-(trifluoromethyl)phenol
SMILES OC1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 122°C
Quantity 1 g
Formula Weight 196.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-hydroxybenzotrifluoride
RTECSNumber SK6040000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.