Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitro-1,4-phenylenediamine 95.0+%, TCI America™
SDP

Catalog No. C150125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C150125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C150125G Supplier TCI America Supplier No. C150125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26196-45-2
Molecular Formula C6H6ClN3O2
Molecular Weight (g/mol) 187.58
MDL Number MFCD00190136
InChI Key VUNAQOGRLGNALG-UHFFFAOYSA-N
Synonym 1,4-Diamino-2-chloro-5-nitrobenzene
PubChem CID 5484290
IUPAC Name 2-chloro-5-nitrobenzene-1,4-diamine
SMILES NC1=CC(=C(N)C=C1Cl)[N+]([O-])=O

Specifications

Melting Point 156°C
Quantity 25 g
Formula Weight 187.58
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitro-1,4-phenylenediamine
RTECSNumber CZ1580000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.