Learn More
Medchemexpress LLC 2-chloro-5-nitro-N-pyridin-4-ylbenzamide | 00-00-0 | MFCD00121849 | 98.1% | C12H4D4ClN3O3 | 10MG
SDP
Catalog No. 5000260690
Encompass_Preferred
T0070907-d4 is the deuterium-labeled analogue of T0070907, a potent and selective peroxisome proliferator-activated receptor gamma (PPARγ) antagonist supplied as a research-grade small molecule. It is intended for biochemical and cellular studies and as an isotopically labeled tracer or internal standard for analytical workflows.
  • Deuterium-labeled analog for tracer and internal standard applications.
  • Potent PPARγ antagonist for pathway and mechanism studies.
  • High purity suitable for analytical and preparative workflows.
  • Stable isotope labeling improves mass spectrometry quantification.
  • Supplied in small research quantities for laboratory use.
  • For research use only; not for clinical or diagnostic use.
Catalog No. 50-002-60690 Supplier Medchemexpress LLC Supplier No. HY13202S10MG
T0070907-d4 is the deuterium-labeled analogue of T0070907, a potent and selective peroxisome proliferator-activated receptor gamma (PPARγ) antagonist supplied as a research-grade small molecule. It is intended for biochemical and cellular studies and as an isotopically labeled tracer or internal standard for analytical workflows.
  • Deuterium-labeled analog for tracer and internal standard applications.
  • Potent PPARγ antagonist for pathway and mechanism studies.
  • High purity suitable for analytical and preparative workflows.
  • Stable isotope labeling improves mass spectrometry quantification.
  • Supplied in small research quantities for laboratory use.
  • For research use only; not for clinical or diagnostic use.