Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-5-nitro-N-pyridin-4-ylbenzamide (d4) | 00-00-0 | MFCD00121849 | 281.69 | C12H4D4ClN3O3 | 5 MG
SDP

Catalog No. 5000262198
Encompass_Preferred

T0070907-d4 is the deuterium-labeled isotopologue of a selective PPARγ antagonist intended for research applications. It is used as an analytical or tracer standard and in mechanistic studies where isotope labeling improves detection, quantitation, or metabolic tracing.

  • Deuterium-labeled isotopologue (d4) for tracer and analytical studies.
  • Potent PPARγ antagonist suitable for receptor biology assays.
  • Molecular formula C12H4D4ClN3O3; molecular weight 281.69.
  • Available in small analytical quantities (e.g., 1 mg, 5 mg, 10 mg).
  • Storage: powder -20°C; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-62198 Supplier Medchemexpress LLC Supplier No. HY13202S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T0070907-d4 is the deuterium-labeled isotopologue of a selective PPARγ antagonist intended for research applications. It is used as an analytical or tracer standard and in mechanistic studies where isotope labeling improves detection, quantitation, or metabolic tracing.

  • Deuterium-labeled isotopologue (d4) for tracer and analytical studies.
  • Potent PPARγ antagonist suitable for receptor biology assays.
  • Molecular formula C12H4D4ClN3O3; molecular weight 281.69.
  • Available in small analytical quantities (e.g., 1 mg, 5 mg, 10 mg).
  • Storage: powder -20°C; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.