Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-5-nitrobenzene sulfonamide | 96-72-0 | 236.64 | 100 G
SDP

Catalog No. 5000449400
Encompass_Preferred

2-Chloro-5-nitrobenzene sulfonamide is a biochemical reagent intended for research use only.

  • Molecular weight: 236.64
  • Chemical formula: C6H5ClN2O4S
  • CAS number: 96-72-0
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-49400 Supplier Medchemexpress LLC Supplier No. HYW008497100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-5-nitrobenzene sulfonamide is a biochemical reagent intended for research use only.

  • Molecular weight: 236.64
  • Chemical formula: C6H5ClN2O4S
  • CAS number: 96-72-0
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.