Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrobenzoic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018322 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8322 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8322 Supplier MilliporeSigma Supelco Supplier No. PHR1741200MG
Add to Cart
Add to Cart
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 2516-96-3
Linear Formula ClC6H3(NO2)CO2 H
MDL Number MFCD00007294
Melting Point 165°C to 168°C
Synonym 2-Chloro-5-nitrobenzoic acid
Type Certified Reference Material
Quantity 200 mg
Formula Weight 201.56
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.