Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrobenzonitrile, 99%

Catalog No. p-7027495
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2428706 5 g
AAB2428714 25 g
AAB2428722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2428706 Supplier Thermo Scientific Chemicals Supplier No. B2428706
Only null left
Add to Cart

CAS: 16588-02-6 | C7H3ClN2O2 | 182.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16588-02-6
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00007292
InChI Key ZGILLTVEEBNDOB-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro-5-nitro, 2-chlor-5-nitrobenzonitril german, 5-nitro-2-chlorobenzonitrile, chembl52706, 2-chloro-5-nitro-benzonitrile, 2-chloro-5-nitrobenzenecarbonitrile, 4-chloro-3-cyano-1-nitrobenzene, 2-chlor-5-nitrobenzonitril, pubchem4740, 2-chloro-5-nitrobenzonitril
PubChem CID 85504
IUPAC Name 2-chloro-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl

Specifications

Melting Point 104°C to 107°C
Quantity 5 g
Beilstein 1818642
Formula Weight 182.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-nitrobenzonitrile
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-643-6
RTECSNumber DI3040000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.