Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrocinnamic Acid 97.0+%, TCI America™
SDP

Catalog No. C12065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12065G 5 g
C120625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12065G Supplier TCI America Supplier No. C12065G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36015-19-7
Molecular Formula C9H5ClNO4
Molecular Weight (g/mol) 226.59
MDL Number MFCD00063312
InChI Key IHTLKLSRRSWIGD-DAFODLJHSA-M
Synonym 2-chloro-5-nitrocinnamic acid, 2e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 3-2-chloro-5-nitrophenyl acrylic acid, 3-2-chloro-5-nitrophenyl propenoic acid, 2-chloro-5-nitrocinnamic acid, predominantly trans, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid, e-3-2-chloro-5-nitrophenyl prop-2-enoic acid, 2e-3-2-chloro-5-nitrophenyl-2-propenoic acid #
PubChem CID 6921604
IUPAC Name (2E)-3-(2-chloro-5-nitrophenyl)prop-2-enoate
SMILES [O-]C(=O)\C=C\C1=CC(=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 221°C
Color Yellow
Quantity 5 g
Formula Weight 227.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrocinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.