Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)aniline, 97%

Catalog No. AAA1600422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1600422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1600422 Supplier Thermo Scientific Chemicals Supplier No. A1600422
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 121-50-6 | C7H5ClF3N | 195.569 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-50-6
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.569
MDL Number MFCD00007673
InChI Key VKTTYIXIDXWHKW-UHFFFAOYSA-N
Synonym 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda
PubChem CID 8475
IUPAC Name 2-chloro-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)Cl

Specifications

Melting Point 8°C to 10°C
Density 1.424
Boiling Point 82°C to 83°C (9 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.499
Quantity 100 g
UN Number UN2810
Beilstein 879910
Formula Weight 195.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)aniline
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 204-475-7
RTECSNumber XU5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.