Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Catalog No. AAB2444906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2444906 5 g
AAB2444903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2444906 Supplier Thermo Scientific Chemicals Supplier No. B2444906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15996-78-8
Molecular Formula C8H8ClF3N
Molecular Weight (g/mol) 210.60
MDL Number MFCD00190123
InChI Key XOCMZYUQWVACMA-UHFFFAOYSA-O
Synonym 2-chloro-5-trifluoromethyl benzylamine, 2-chloro-5-trifluoromethyl phenyl methanamine, 2-chloro-5-trifluoromethyl-benzylamine, 2-chloro-5-trifluoromethylbenzylamine, 2-chloro-5-trifluoromethylbenzyl amine, 2-chloro-5-trifluoromethyl benzyl amine, 2-chloro-5-trifluoromethyl phenyl methylamine, 1-2-chloro-5-trifluoromethyl phenyl methanamine, pubchem4392, rarechem al bw 0385
PubChem CID 2736682
SMILES [NH3+]CC1=CC(=CC=C1Cl)C(F)(F)F

Specifications

Boiling Point 90°C to 94°C (13 mmHg)
Flash Point 100°C (212°F)
Odor Amine-like
Quantity 5 g
UN Number UN2735
Beilstein 2835666
IUPAC Name [2-chloro-5-(trifluoromethyl)phenyl]methanamine
Formula Weight 209.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzylamine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.