Learn More
2-Chloro-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 90°C to 94°C (13 mmHg) |
| Flash Point | 100°C (212°F) |
| Odor | Amine-like |
| Quantity | 1 g |
| UN Number | UN2735 |
| Beilstein | 2835666 |
| IUPAC Name | [2-chloro-5-(trifluoromethyl)phenyl]methanaminium |
| Formula Weight | 209.6 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-5-(trifluoromethyl)benzylamine |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement P280-P305+P351+P338-P309-P310 Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician. |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.