Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Catalog No. AAH63350MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63350MD 250 mg
AAH6335003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63350MD Supplier Thermo Scientific Chemicals Supplier No. H63350MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505084-59-3
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD08741353
InChI Key RMSKYTKOFDQVEL-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl nicotinic acid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-5-trifluoromethyl-3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethylnicotinic acid, 2-chloro-5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, ksc269c4j, 2-chloro-5-trifluoromethyl nicotinicacid, 2-chloro-5-trifluoromethyl pyridine-3-carboxylic, 2-chloro-5-trifluoromethyl pyridine-3-carboxylicacid
PubChem CID 12140227
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=C(C(=NC=C1C(F)(F)F)Cl)C(=O)O

Specifications

Melting Point 173°C to 178°C
Quantity 250 mg
Formula Weight 225.55
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)nicotinic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.