Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)pyrazine, 97%

Catalog No. AC446480050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446480050 5 g
AC446480010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446480050 Supplier Thermo Scientific Chemicals Supplier No. 446480050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 799557-87-2 | C5H2ClF3N2 | 182.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 799557-87-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
InChI Key AIEGIFIEQXZBCP-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine
PubChem CID 18987645
IUPAC Name 2-chloro-5-(trifluoromethyl)pyrazine
SMILES C1=C(N=CC(=N1)Cl)C(F)(F)F

Specifications

Boiling Point 88°C (118.0 mmHg)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 182.53
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyrazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.