Learn More
Medchemexpress LLC 2-chloro-6-fluorobenzaldehyde | 387-45-1 | MFCD00003306 | 99.7% | 158.56 g/mol | C7H4ClFO | 1 KG
SDP
Catalog No. 5000302792
Encompass_Preferred
2-Chloro-6-fluorobenzaldehyde is an aromatic aldehyde used as a synthetic building block in organic chemistry and research. It is employed in condensation reactions and other transformations to introduce a 2-chloro-6-fluorophenyl-CHO motif; consult the batch COA for exact specifications.
  • High reported purity; verify batch COA for exact value.
  • Available in multiple sizes including kilo-scale for scale-up work.
  • Supplied with certificate of analysis and safety data sheet.
  • Solid at ambient temperatures below ~32°C; may liquefy above ~35°C.
  • Useful building block for pharmaceutical and agrochemical synthesis.
Catalog No. 50-003-02792 Supplier Medchemexpress LLC Supplier No. HYW0154771KG
2-Chloro-6-fluorobenzaldehyde is an aromatic aldehyde used as a synthetic building block in organic chemistry and research. It is employed in condensation reactions and other transformations to introduce a 2-chloro-6-fluorophenyl-CHO motif; consult the batch COA for exact specifications.
  • High reported purity; verify batch COA for exact value.
  • Available in multiple sizes including kilo-scale for scale-up work.
  • Supplied with certificate of analysis and safety data sheet.
  • Solid at ambient temperatures below ~32°C; may liquefy above ~35°C.
  • Useful building block for pharmaceutical and agrochemical synthesis.