Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-6-fluorobenzaldehyde | 387-45-1 | MFCD00003306 | 99.7% | 158.56 g/mol | C7H4ClFO | 100 G
SDP

Catalog No. 5000302532
Encompass_Preferred

2-Chloro-6-fluorobenzaldehyde is a halogenated benzaldehyde used as an intermediate in organic synthesis and as a reagent for chemical research. The material is supplied with a certificate of analysis documenting purity, identity, and storage recommendations to ensure reproducible results and stability.

  • High purity: 99.69% (GC), suitable for synthetic applications.
  • Molecular weight: 158.56 g/mol.
  • Chemical formula: C7H4ClFO.
  • Appearance: White to off-white solid with melting/appearance notes.
  • Storage: Recommend 4°C under inert gas; in solvent store at -80°C for long term.

Catalog No. 50-003-02532 Supplier Medchemexpress LLC Supplier No. HYW015477100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloro-6-fluorobenzaldehyde is a halogenated benzaldehyde used as an intermediate in organic synthesis and as a reagent for chemical research. The material is supplied with a certificate of analysis documenting purity, identity, and storage recommendations to ensure reproducible results and stability.

  • High purity: 99.69% (GC), suitable for synthetic applications.
  • Molecular weight: 158.56 g/mol.
  • Chemical formula: C7H4ClFO.
  • Appearance: White to off-white solid with melting/appearance notes.
  • Storage: Recommend 4°C under inert gas; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.