Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzoic acid, 98+%, Thermo Scientific™

Catalog No. AAA1125006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1125006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1125006 Supplier Thermo Scientific Chemicals Supplier No. A1125006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-75-3
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00002417
InChI Key XNTIGDVFBDJLTQ-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-6-fluoro, unii-euq5bp41s0, euq5bp41s0, 2-fluoro-6-chlorobenzoic acid, gnf-pf-2650, 2-chloro-6-fluoro-benzoic acid, 2-chloro-6-fluorobenzoicacid, pubchem1372, acmc-1ctwn, ksc238i3p
PubChem CID 67947
IUPAC Name 2-chloro-6-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)F

Specifications

Melting Point 156°C to 159°C
Quantity 5 g
Beilstein 973857
Formula Weight 174.56
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-6-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-105-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.