Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. C230825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C230825G 25 g
C23085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C230825G Supplier TCI America Supplier No. C230825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68220-26-8
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD00040126
InChI Key IGUVNNXFTDCASP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl bromide, 2-bromomethyl-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzylbromide, 2-bromomethyl-1-chloro-3-fluoro-benzene, benzene, 2-bromomethyl-1-chloro-3-fluoro, 2-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-2-chloro-6-fluorotoluene, pubchem4894, acmc-1bdpq
PubChem CID 2773625
IUPAC Name 2-(bromomethyl)-1-chloro-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1CBr

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 223.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-fluorobenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.