Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C108125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C108125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C108125G Supplier TCI America Supplier No. C108125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55117-15-2
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.02
MDL Number MFCD00000896
InChI Key MJGOLNNLNQQIHR-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl chloride, 1-chloro-2-chloromethyl-3-fluorobenzene, benzene, 1-chloro-2-chloromethyl-3-fluoro, 2-chloro-6-fluorobenzylchloride, unii-a581kl6zpn, 1-chloro-2-chloromethyl-3-fluoro-benzene, alpha,2-dichloro-6-fluorotoluene, a581kl6zpn, 3-chloro-2-chloromethyl-1-fluorobenzene, pubchem7497
PubChem CID 108675
IUPAC Name 1-chloro-2-(chloromethyl)-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1CCl

Specifications

Color Colorless
Boiling Point 210°C
Quantity 25 g
UN Number 3265
Formula Weight 179.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-fluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.