Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. C20495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20495G 5 g
C204925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20495G Supplier TCI America Supplier No. C20495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2040-90-6
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
InChI Key QIAQIYQASAWZPP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-phenol, 2-chloro-6-fluoro phenol, phenol, 2-chloro-6-fluoro, pubchem2853, 6-chlor-2-fluorphenol, acmc-1cqou, 6-chloro-2-fluorophenol, 2-chlor-6-fluorphenol;, 2-chloro-6-fluorophenol;, 2-chlor-6-fluor-phenol;
PubChem CID 2773710
IUPAC Name 2-chloro-6-fluorophenol
SMILES C1=CC(=C(C(=C1)Cl)O)F

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 146.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.