Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C32471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32471G 1 g
C3247-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32471G Supplier TCI America Supplier No. C32471G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 2-Chloro-6-fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 313545-32-3
Melting Point 135°C
Color White-Yellow
Molecular Formula C6H5BClFO2
Quantity 1 g
MDL Number MFCD04039892
Synonym 2-chloro-6-fluorophenyl boronic acid, 2-chloro-6-fluorobenzeneboronic acid, 2-chloro-6-fluorophenylboronicacid, boronic acid, 2-chloro-6-fluorophenyl, boronic acid,b-2-chloro-6-fluorophenyl, boronic acid, b-2-chloro-6-fluorophenyl, pubchem18883, acmc-1ad0v, 2-borono-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzene boronic acid
InChI Key NXSZSZJWZVLHAY-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C=CC=C1Cl
IUPAC Name (2-chloro-6-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.36
PubChem CID 3549395
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.