Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxy-3-nitropyridine, 98%

Catalog No. AAH6407622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6407622 100 g
AAH6407614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6407622 Supplier Thermo Scientific Chemicals Supplier No. H6407622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 38533-61-8 | C6H5ClN2O3 | 188.57 g/mol

6-methoxy-3-nitropyridine-2-carbonitrile was synthesised from 2-Chloro-6-methoxy-3-nitropyridine. Reatant in the suzuki and negishi couplings reaction. Also as fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-methoxy-3-nitropyridine-2-carbonitrile was synthesised from 2-Chloro-6-methoxy-3-nitropyridine. Reatant in the suzuki and negishi couplings reaction. Also as fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38533-61-8
Molecular Formula C6H5ClN2O3
Molecular Weight (g/mol) 188.57
MDL Number MFCD00130268
InChI Key DVRGUTNVDGIKTP-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine
PubChem CID 2795029
IUPAC Name 2-chloro-6-methoxy-3-nitropyridine
SMILES COC1=CC=C(C(Cl)=N1)[N+]([O-])=O

Specifications

Melting Point 78°C to 80°C (literature)
Color White to Yellow
Quantity 100 g
UN Number UN2811
Beilstein 1454907
Solubility Information Slightly soluble in water.
Formula Weight 188.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-6-methoxy-3-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 253-989-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.