Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxy-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. C16365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16365G 5 g
C163625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16365G Supplier TCI America Supplier No. C16365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38533-61-8
Molecular Formula C6H5ClN2O3
Molecular Weight (g/mol) 188.57
MDL Number MFCD00130268
InChI Key DVRGUTNVDGIKTP-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine
PubChem CID 2795029
IUPAC Name 2-chloro-6-methoxy-3-nitropyridine
SMILES COC1=CC=C(C(Cl)=N1)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 188.57
Percent Purity ≥98.0% (GC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methoxy-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.