Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-methoxyisonicotinic acid, 97%, Thermo Scientific™

Catalog No. RF03693EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RF03693EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF03693EA Supplier Thermo Scientific Chemicals Supplier No. RF03693EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15855-06-8
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00173928
InChI Key PJQBTHQTVJMCFX-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxyisonicotinic acid, 2-chloro-6-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy, 2-chloro-6-methoxy-4-pyridinecarboxylic acid, 2-chloro-6-methoxyisonicotinicacid, acmc-1bs54, ksc173i0h, 2-chloro-6-methoxypyridine-4-carboxylicacid
PubChem CID 2782124
IUPAC Name 2-chloro-6-methoxypyridine-4-carboxylic acid
SMILES COC1=NC(=CC(=C1)C(=O)O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 187.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methoxyisonicotinic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.