Learn More
Medchemexpress LLC 2-Chloro-6-methoxypyridine | 17228-64-7 | 143.57 | 25 G
SDP
Catalog No. 5000448260
Encompass_Preferred
2-Chloro-6-methoxypyridine is a biochemical reagent used for life science-related research. It is intended for research use only and not for patient use.
  • Used as a biological material or organic compound
  • Suitable for life science research
  • Liquid appearance
  • Colorless to light yellow liquid
  • Pure form stable for 3 years at -20°C
  • Pure form stable for 2 years at 4°C
  • In solvent stable for 6 months at -80°C
  • In solvent stable for 1 month at -20°C
Catalog No. 50-004-48260 Supplier Medchemexpress LLC Supplier No. 25GHYW001995
2-Chloro-6-methoxypyridine is a biochemical reagent used for life science-related research. It is intended for research use only and not for patient use.
  • Used as a biological material or organic compound
  • Suitable for life science research
  • Liquid appearance
  • Colorless to light yellow liquid
  • Pure form stable for 3 years at -20°C
  • Pure form stable for 2 years at 4°C
  • In solvent stable for 6 months at -80°C
  • In solvent stable for 1 month at -20°C