Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxypyridine-4-carbonyl chloride, 95%, Thermo Scientific™

Catalog No. AAH5015906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5015906 5 g
AAH5015903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5015906 Supplier Thermo Scientific Chemicals Supplier No. H5015906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116853-97-5
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
MDL Number MFCD00125355
InChI Key RITCMXVSFUDYGY-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxyisonicotinoyl chloride, 2-chloro-6-methoxypyridine-4-carbonylchloride, zlchem 998, pubchem17742, acmc-20a3t1, 2-chloro-4-chlorocarbonyl-6-methoxypyridine, 2-chloro-6-methoxy-4-pyridinecarbonyl chloride, 2-chloro-6-methoxy-pyridine-4-carbonyl chloride, 4-pyridinecarbonylchloride, 2-chloro-6-methoxy, 2-chloranyl-6-methoxy-pyridine-4-carbonyl chloride
PubChem CID 2782125
IUPAC Name 2-chloro-6-methoxypyridine-4-carbonyl chloride
SMILES COC1=NC(=CC(=C1)C(=O)Cl)Cl

Specifications

Boiling Point 79°C (0.2 mmHg)
Refractive Index 1.556
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 206.03
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methoxypyridine-4-carbonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.