Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylaniline, Thermo Scientific™

Catalog No. AC109360250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109360250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC109360250 Supplier Thermo Scientific Chemicals Supplier No. 109360250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007679
InChI Key WFNLHDJJZSJARK-UHFFFAOYSA-N
Synonym 6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
PubChem CID 6897
IUPAC Name 2-chloro-6-methylaniline
SMILES CC1=CC=CC(Cl)=C1N

Specifications

Melting Point 2.0°C
Density 1.1520g/mL
Boiling Point 215.0°C
Flash Point 98°C
Refractive Index 1.5750 to 1.5780
Linear Formula ClC6H3(CH3)NH2
Quantity 25 mL
Solubility Information Solubility in water: immiscible in water. Other solubilities: miscible with toluene and chlorobenzene
Formula Weight 141.6
Chemical Name or Material 2-Chloro-6-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.