Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylbenzoic acid, 97%

Catalog No. AAL1834806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1834806 5 g
AAL1834803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1834806 Supplier Thermo Scientific Chemicals Supplier No. L1834806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 21327-86-6 | C8H7ClO2 | 170.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21327-86-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045799
InChI Key CEFMMQYDPGCYMG-UHFFFAOYSA-N
Synonym 6-chloro-o-toluic acid, 2-chloro-6-methyl-benzoic acid, o-toluic acid, 6-chloro, 2-chloro-6-methylbenzoicacid, benzoic acid, 2-chloro-6-methyl, benzoic acid, 6-chloro-4-methyl, 6-chloro-2-methylbenzoic acid, pubchem4568, chloro-6-methylbenzoic acid, 2-m-tolyloxymethyl oxirane
PubChem CID 88870
IUPAC Name 2-chloro-6-methylbenzoic acid
SMILES CC1=C(C(=CC=C1)Cl)C(=O)O

Specifications

Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2614092
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber XU1645100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.