Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. C31605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31605G 5 g
C31601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31605G Supplier TCI America Supplier No. C31605G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3507-26-4
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD04448821
InChI Key PAKSGYIFUVNJQF-UHFFFAOYSA-N
PubChem CID 2049866
IUPAC Name 2-chloro-6-methyl-1,3-benzothiazole
SMILES CC1=CC2=C(C=C1)N=C(S2)Cl

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 183.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.