Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylisonicotinic acid, 95+%, Thermo Scientific™

Catalog No. RF03759EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF03759EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF03759EA Supplier Thermo Scientific Chemicals Supplier No. RF03759EA
Add to Cart

Chemical Identifiers

CAS 25462-85-5
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
InChI Key ZGZMEKHQIZSZOH-UHFFFAOYSA-N
Synonym 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
PubChem CID 141209
IUPAC Name 2-chloro-6-methylpyridine-4-carboxylic acid
SMILES CC1=NC(=CC(=C1)C(=O)O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 171.58
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.