Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methylisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. C24861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24861G 1 g
C24865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24861G Supplier TCI America Supplier No. C24861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25462-85-5
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00052830
InChI Key ZGZMEKHQIZSZOH-UHFFFAOYSA-N
Synonym 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
PubChem CID 141209
IUPAC Name 2-chloro-6-methylpyridine-4-carboxylic acid
SMILES CC1=NC(=CC(=C1)C(=O)O)Cl

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 1 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.