Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-nitrobenzaldehyde, MP Biomedicals

Catalog No. ICN224002 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN224002 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN224002 Supplier MP Biomedicals, Inc Supplier No. 0522400201

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6361-22-4
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007204
InChI Key RZDOUWDCYULHJX-UHFFFAOYSA-N
Synonym 2-nitro-6-chlorobenzaldehyde, benzaldehyde, 2-chloro-6-nitro, 6-chloro-2-nitrobenzaldehyde, pubchem17009, 2-chloro-6-nitro-benzaldehye, ksc497q1l, 2-chloro-6-nitro-benzaldehyde, benzaldehyde,2-chloro-6-nitro, rzdouwdcyulhjx-uhfffaoysa, attercop-chm at114429
PubChem CID 80701
IUPAC Name 2-chloro-6-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=CC(Cl)=C1C=O

Specifications

Quantity 1 g
Formula Weight 185.57
Chemical Name or Material 2-Chloro-6-nitrobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.