Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-epsilon-caprolactone

Catalog No. 9008781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-087-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-087-81G Supplier Sigma Aldrich Supplier No. 9008781G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloro-α-caprolactone (ClCL) is a biomaterial that can be prepared by the Baeyer-Villiger oxidation of α-chlorocyclohexanone. It can be copolymerized with α-caprolactone for the formation of Poly(2-chloro-α-caprolactone).2-Chloro-epsilon-caprolactone (or alpha-Chloro-epsilon-caprolactone) is a functionalized biodegradable monomer. This monomer can be polymerized using ring-opening polymerization to yield a chloride-functionalized polymer backbone that can either be further functionalized with small molecules or used in the synthesis of graft co-polymers.

Specifications

CAS 89727-99-1
Molecular Formula C6H9ClO2
Linear Formula C6H9ClO2
MDL Number MFCD20622420
Quantity 1 g
Synonym alpha-Chloro-epsilon-caprolactone; alphaClepsilonCL; 2-Chloro-1-oxacycloheptan-2-one
Molecular Weight (g/mol) 148.59
Percent Purity 98% (NMR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.