Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-N-[2-(4-chlorophenyl)phenyl]pyridine-3-carboxamide | 188425-85-6 | 99.8% | 1 ML
SDP

Catalog No. 5000234085
Encompass_Preferred

Boscalid is an antifungal agent and a succinate dehydrogenase (SDH) inhibitor supplied as a 10 mM solution (1 mL) for research use.

  • Antifungal agent and SDH inhibitor.
  • Provided as a 10 mM solution, 1 mL.
  • High purity for research applications (COA-supported).
  • Molecular weight 343.21 g/mol; formula C18H12Cl2N2O.
  • Soluble in DMSO; suitable for in vitro assay preparation.
  • Intended for research use only.

Catalog No. 50-002-34085 Supplier Medchemexpress LLC Supplier No. HY11997610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boscalid is an antifungal agent and a succinate dehydrogenase (SDH) inhibitor supplied as a 10 mM solution (1 mL) for research use.

  • Antifungal agent and SDH inhibitor.
  • Provided as a 10 mM solution, 1 mL.
  • High purity for research applications (COA-supported).
  • Molecular weight 343.21 g/mol; formula C18H12Cl2N2O.
  • Soluble in DMSO; suitable for in vitro assay preparation.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.