Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloro-N-[2-[[4-[methyl(4-phenylbutyl)amino]-6-(prop-2-ynylamino)-1,3,5-triazin-2-yl]amino]ethyl]acetamide | 2226201-97-2 | 99.9% | 429.95 g/mol | C21H28ClN7O | 50 MG
SDP

Catalog No. 5000226771
Encompass_Preferred

KSC-34 is a covalent modifier and selective inhibitor of protein disulfide isomerase A1 (PDIA1) used as a research reagent for biochemical and cell-based studies. It is supplied as a solid and intended for laboratory research use only; follow appropriate safety and storage guidelines.

  • Selective PDIA1 a-site inhibitor with IC50 ≈3.5 μM.
  • High purity (≈99.9%).
  • Molecular weight 429.95 g/mol; formula C21H28ClN7O.
  • White to off-white solid, packaged as 50 mg.
  • Soluble in DMSO (100 mg/mL); in vivo formulation options documented.
  • Storage: store at 4°C under nitrogen; in solution, store at -80°C for long-term preservation.

Catalog No. 50-002-26771 Supplier Medchemexpress LLC Supplier No. HY11757050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KSC-34 is a covalent modifier and selective inhibitor of protein disulfide isomerase A1 (PDIA1) used as a research reagent for biochemical and cell-based studies. It is supplied as a solid and intended for laboratory research use only; follow appropriate safety and storage guidelines.

  • Selective PDIA1 a-site inhibitor with IC50 ≈3.5 μM.
  • High purity (≈99.9%).
  • Molecular weight 429.95 g/mol; formula C21H28ClN7O.
  • White to off-white solid, packaged as 50 mg.
  • Soluble in DMSO (100 mg/mL); in vivo formulation options documented.
  • Storage: store at 4°C under nitrogen; in solution, store at -80°C for long-term preservation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.