Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloro-N-(4'-chloro[1,1'-biphenyl]-2-yl)-3-pyridinecarboxamide | 188425-85-6 | 99.8% | 500 MG
SDP

Catalog No. 5000233956
Encompass_Preferred

Boscalid is a broad-spectrum carboxamide fungicide and succinate dehydrogenase (SDH) inhibitor supplied as a solid analytical standard for research and analytical applications. Typical pack contains 500 MG with a listed purity of 99.8%.

  • High purity of 99.8% suitable for analytical use.
  • Formula C18H12Cl2N2O with molecular weight 343.21 g·mol⁻¹.
  • Solid powder appearance for easy handling and formulation.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Mode of action: succinate dehydrogenase (SDH) inhibitor useful in fungal respiration studies.
  • Intended for research and analytical applications; not for human or veterinary use.

Catalog No. 50-002-33956 Supplier Medchemexpress LLC Supplier No. HY119976500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boscalid is a broad-spectrum carboxamide fungicide and succinate dehydrogenase (SDH) inhibitor supplied as a solid analytical standard for research and analytical applications. Typical pack contains 500 MG with a listed purity of 99.8%.

  • High purity of 99.8% suitable for analytical use.
  • Formula C18H12Cl2N2O with molecular weight 343.21 g·mol⁻¹.
  • Solid powder appearance for easy handling and formulation.
  • Storage: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (6 months) or -20°C (1 month).
  • Mode of action: succinate dehydrogenase (SDH) inhibitor useful in fungal respiration studies.
  • Intended for research and analytical applications; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.