Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloro-N-(4-chloro-2-methoxy-5-methylphenyl)acetamide

Catalog No. OTV000026 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000026 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000026 Supplier Sigma Aldrich Supplier No. OTV000026250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl isonipecotate can be obtained by the reduction of methyl isonicotinate in methanol with ruthenium at 100-150C.Reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activationReactant for synthesis of: Antitubercular agents Anti-HIV-1 agents Aminopyrazine inhibitors Orally available naphthyridine protein kinase D inhibitors A Scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide

Specifications

CAS 379255-21-7
Molecular Formula C10H11Cl2NO2
Linear Formula C10H11Cl2NO2
Quantity 25 mL
MDL Number MFCD02965566
Molecular Weight (g/mol) 248.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.