Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-methoxy-N-methylacetamide 98.0+%, TCI America™
SDP

Catalog No. C22725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22725G 5 g
C227225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22725G Supplier TCI America Supplier No. C22725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67442-07-3
Molecular Formula C4H8ClNO2
Molecular Weight (g/mol) 137.563
MDL Number MFCD00134232
InChI Key SCOJKGRNQDKFRP-UHFFFAOYSA-N
PubChem CID 2734716
IUPAC Name 2-chloro-N-methoxy-N-methylacetamide
SMILES CN(C(=O)CCl)OC

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 95°C
Quantity 5 g
Synonym 2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide
Formula Weight 137.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-N-methoxy-N-methylacetamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.