Learn More
Medchemexpress LLC 2-Chloro-N-(methylsulfonyl)acetamide | 202658-88-6 | MFCD20637131 | 98% | C3H6ClNO3S | 10 G
SDP
Catalog No. 5000432870
Encompass_Preferred
2-Chloro-N-(methylsulfonyl)acetamide is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It also functions as a reagent for preparing piperazinylquinolonecarboxamides, which are used as antivirals.
  • Drug intermediate for synthesis of active compounds
  • Reagent for preparing piperazinylquinolonecarboxamides
  • Used in antiviral synthesis
Catalog No. 50-004-32870 Supplier Medchemexpress LLC Supplier No. HY7968110G
2-Chloro-N-(methylsulfonyl)acetamide is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds. It also functions as a reagent for preparing piperazinylquinolonecarboxamides, which are used as antivirals.
  • Drug intermediate for synthesis of active compounds
  • Reagent for preparing piperazinylquinolonecarboxamides
  • Used in antiviral synthesis