Learn More
Cayman Chemical 2-chloro Palmitic Acid, Purity: ≥95%, CAS: 19117-92-1, 1 mg
SDP
Catalog No. 502936721
Encompass_Preferred
A monochlorinated form of palmitic acid; produced in an MPO- and time-dependent manner in neutrophils stimulated by PMA; induces NETosis in human neutrophils, increasing DNA release from neutrophils, colocalization of MPO with ecDNA, and trapping of E. coli at 10 μM; increases COX-2 protein levels in HCAECs at 50 μM; increases production of P-selectin, von Willebrand factor, and angiopoietin-2 in HCAECs, as well as neutrophil and platelet adherence, at 10 μM; induces apoptosis in THP-1 cells and primary human monocytes and increases caspase-3 activity in THP-1 cells at 10-50 μM
Catalog No. 50-293-6721 Supplier Cayman Chemical Supplier No. 261071 MG
A monochlorinated form of palmitic acid; produced in an MPO- and time-dependent manner in neutrophils stimulated by PMA; induces NETosis in human neutrophils, increasing DNA release from neutrophils, colocalization of MPO with ecDNA, and trapping of E. coli at 10 μM; increases COX-2 protein levels in HCAECs at 50 μM; increases production of P-selectin, von Willebrand factor, and angiopoietin-2 in HCAECs, as well as neutrophil and platelet adherence, at 10 μM; induces apoptosis in THP-1 cells and primary human monocytes and increases caspase-3 activity in THP-1 cells at 10-50 μM