Learn More
Cayman Chemical 2-chloro Stearic Acid, Purity: ≥95%, CAS: 56279-49-3, 1 mg
SDP
Catalog No. 502934340
Encompass_Preferred
A bioactive fatty acid; accumulates in PMA-stimulated primary human monocytes and neutrophils as well as murine neutrophils; induces DNA release from primary human neutrophils
Catalog No. 50-293-4340 Supplier Cayman Chemical Supplier No. 261081 MG
A bioactive fatty acid; accumulates in PMA-stimulated primary human monocytes and neutrophils as well as murine neutrophils; induces DNA release from primary human neutrophils