Learn More
Selleck Chemical LLC 2-Chloroacetamide E2952-25MG
SDP
Catalog No. 5000040579
Encompass_Preferred
2-Chloroacetamide is a preservative and is a herbicide for both uplands and paddy fields 2-Chloroacetamide is a biocide in agriculture glues paints and coatings 2-Chloroacetamide inhibits very-long-chain fatty acid elongase
Catalog No. 50-000-40579 Supplier Selleck Chemical LLC Supplier No. E295225MG
2-Chloroacetamide is a preservative and is a herbicide for both uplands and paddy fields 2-Chloroacetamide is a biocide in agriculture glues paints and coatings 2-Chloroacetamide inhibits very-long-chain fatty acid elongase