Learn More
Medchemexpress LLC 2-Chloroacetamide | 79-07-2 | 99.9% | 93.51 | 1 ML
SDP
Catalog No. 5000453605
Encompass_Preferred
2-Chloroacetamide is a preservative and a herbicide for both uplands and paddy fields. It acts as a biocide in agriculture, glues, paints, and coatings. It also inhibits very-long-chain fatty acid elongase.
  • Preservative
  • Herbicide for uplands and paddy fields
  • Biocide in agriculture, glues, paints, and coatings
  • Inhibits very-long-chain fatty acid elongase
  • Herbicide for paddy field weeds, such as barnyard grass
Catalog No. 50-004-53605 Supplier Medchemexpress LLC Supplier No. HYW01062910MM/1ML
2-Chloroacetamide is a preservative and a herbicide for both uplands and paddy fields. It acts as a biocide in agriculture, glues, paints, and coatings. It also inhibits very-long-chain fatty acid elongase.
  • Preservative
  • Herbicide for uplands and paddy fields
  • Biocide in agriculture, glues, paints, and coatings
  • Inhibits very-long-chain fatty acid elongase
  • Herbicide for paddy field weeds, such as barnyard grass