Learn More
Medchemexpress LLC 2-Chloroacetamide | 79-07-2 | 99.9% | 93.51 | 10 G
SDP
Catalog No. 5000366551
Encompass_Preferred
2-Chloroacetamide is a chemical compound that serves as a preservative and herbicide for both uplands and paddy fields. It is also used as a biocide in agriculture, glues, paints, and coatings. This compound is known for its ability to inhibit very-long-chain fatty acid elongase.
  • Functions as a preservative
  • Functions as a herbicide
  • Acts as a biocide in agriculture, glues, paints, and coatings
  • Inhibits very-long-chain fatty acid elongase
  • Demonstrates herbicidal activities against paddy field weeds
Catalog No. 50-003-66551 Supplier Medchemexpress LLC Supplier No. HYW01062910G
2-Chloroacetamide is a chemical compound that serves as a preservative and herbicide for both uplands and paddy fields. It is also used as a biocide in agriculture, glues, paints, and coatings. This compound is known for its ability to inhibit very-long-chain fatty acid elongase.
  • Functions as a preservative
  • Functions as a herbicide
  • Acts as a biocide in agriculture, glues, paints, and coatings
  • Inhibits very-long-chain fatty acid elongase
  • Demonstrates herbicidal activities against paddy field weeds